Heat Exchangers for Bioreactors, Chemglass
Supplier: Chemglass
CLS-1380-28 CLS-1382-11 CLS-1383-12 CLS-1385-16
10170-870EA 1259.17 CAD
10170-870 10170-872 10170-874 75780-154
Heat Exchangers for Bioreactors, Chemglass
Circulating Thermostats Closed Circulator Systems Heating Circulators
Heat exchangers are available in a choice of two different styles.
The cold finger style is for use with 3 and 5 litre reactors and fits into the headplate via an M18 threaded port. The second style is a loop type exchanger and is available for 5 - 15 litre reactors. It fits into two, M10 ports on the headplate.