OHAUS® Incubating Cooling Thermal Shakers
Supplier: OHAUS CORP.
30391996 30392003 30400126 30400127 30400128 30400129 30400130 30400131 30400250 30400251 30400252 30400134 30400135 30400136
470235-152EA 6163.85 CAD
470235-152 470235-154 470235-170 470235-172 470235-174 470235-176 470235-178 470235-180 470235-182 470235-184 470235-186 470235-188 470235-190 470235-192 470235-194 470235-196
OHAUS® Incubating Cooling Thermal Shakers
Shakers Shaking Incubators
Intuitive Incubating Cooling Thermal Shakers for Consistent High-Speed Shaking.
  • 4.3" color LCD touch screen display provides an intuitive interface
  • Rapid heating, cooling and high speed shaking ability
  • Internal memory stores five separate 5-step programs, unlimited with USB
The OHAUS Thermal Shakers and Cooling Thermal Shakers are designed for applications that require consistent and precise results. With heating, cooling and shaking capabilities. These shakers use interchangeable blocks to accommodate a wide variety of tubes and microplates. The easy-to-use, 4.3", color, LCD touch screen allows the user to save and visibly track progress through the live status bar for five user defined programs, each with five individual steps. The unit’s enhanced electronics and dual temperature sensors provide accurate, dependable temperature settings across the operating range.
Operating Features: Low Profile Design-The low profile design minimizes the unit’s footprint on the bench. LCD Touch Screen-Enables faster setting of temperature, speed, and time which can all be viewed at once. Display features on-screen help topics with operational tips available in six languages. Touch screen is compatible with rubber gloves used in labs. USB port can transfer information to a flash drive for data logging, program storage and software updates. Program Control-Program control capabilities allow user programmable operation for automated use and memory. Storage for five separate 5-step programs, or unlimited number of programs with the use of the USB. Temperature ramp rate- Adjustable temperature ramp rate feature separately defines the heating and cooling rate in increments of 0.5 °C/min. Single Point Calibration Mode- For maximum temperature accuracy, the single point calibration procedure allows the user to calibrate up to 6 different user defined temperatures. Pulse Mode Feature- The unit is equipped with a pulse mode feature for quick vortex applications.
Safety Features: Cool Touch Housing–Constructed from a high-quality, heat and chemical resistant polymer. The unit’s housing remains cool to the touch throughout normal operating temperatures. Maximum Temperature Limiting Function-Ensures the temperature will not exceed preset limits, allowing the user control of temperature sensitive samples. Hot Top Indicator-A hot top warning light will illuminate when the temperature reaches 40 °C, and will remain lit until the unit is sufficiently cooled. Audible Alarm-In timed mode, an alarm will sound when the time reaches zero or set-point temperature is reached. Additionally, the heat function will automatically shut off if the unit recognizes an internal issue.
Operating Conditions–Unit can operate in conditions from 5 to 35 °C (41 to 95 °F), maximum 80% relative humidity, non-condensing. Applications- Cell cultures, DNA, RNA, and protein studies.
Ordering information: Unit includes a detachable, 3-wire cord and plug. Unit is also supplied with a 1.5 mL block, clear rack, and cover.