L-Arginine hydrochloride ≥99% (by titrimetric analysis), Calbiochem®

Supplier: MilliporeSigma

181003-250GM 181003-50GM
CA80108-170EA 402.65 CAD
CA80108-170 CA80108-172
L-Arginine hydrochloride ≥99% (by titrimetric analysis), Calbiochem®
L-Arginine hydrochloride
Formula: C₆H₁₅ClN₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2

Order Now

Specification Test Results

Purity ≥99% (by perchloric acid titration)
Contaminants (iron) ≤10 ppm
Contaminants (endotoxin) ≤6.0 EU/g
Contaminants (total micoorganisms) ≤100 CFU/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR