PfuUltra High-Fidelity DNA Polymerase, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
PfuUltra High-Fidelity DNA Polymerase AD features a reformulated buffer system for increased economy with the same high performance as the original PfuUltra DNA Polymerase for robust, ultra-high-fidelity PCR.
PfuUltra Hotstart DNA Polymerase features a genetically engineered mutant of Pfu DNA Polymerase and the ArchaeMaxx Polymerase Enhancing Factor and is formulated for hot start activity.
PfuUltra Hotstart DNA Polymerase AD features a reformulated buffer system and novel hot start mechanism for increased economy with the same high performance as the original PfuUltra Hotstart DNA Polymerase.
PfuUltra Hotstart PCR Master Mix is a 2X formulation of PfuUltra Hotstart DNA Polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.