AdEasy Adenoviral Vector Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
240071
240073
240109
CAST240071EA
1925.97
CAD
CAST240071
CAST240073
CAST240109
AdEasy Adenoviral Vector Systems, Agilent Technologies
Vectors, Plasmids and Libraries
The AAV helper-free system eliminates the requirement for wild-type adenovirus co-infection from both AAV vector production and AAV stock titering steps, making this system entirely helper virus-free and safer.
- Stable gene expression
- High titer virus
- Lower toxicity vs. non-viral methods of gene delivery
Caution: For research use only. Not for use in diagnostic procedures.