Multiple Affinity Removal Column Human 14, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
5188-6558 5188-6557
CA76193-636EA 10809.42 CAD
CA76193-636 CA76193-736
Multiple Affinity Removal Column Human 14, Agilent Technologies
Chromatography Columns
The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.
  • Made of PEEK (polyetheretherketone)
  • High-capacity serum load
  • Affinity resin packed
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.