StrataClone PCR Cloning Kits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
240218 240205 240207
CA76193-372EA 725.55 CAD
CA76193-372 CA99901-042 CA99901-044
StrataClone PCR Cloning Kits, Agilent Technologies
Nucleic Acid Reagents Cloning Kits and Ligation
The StrataClone PCR Cloning Kit allows high-efficiency, 5-minute cloning of PCR products at room temperature, using the efficient DNA rejoining activity of DNA topoisomerase I and the DNA recombination activity of Cre recombinase.
  • High efficiency yields >95% insert positive clones
  • Clone both long and short amplicons (500 bp to 9.2 Kb)
  • With kanamycin and ampicillin resistance markers
These 3 kits are available for both blunt-end and UA cloning. Additionally, combined the highest accuracy in PCR amplification with easy, reliable, and affordably priced topoisomerase-based cloning in StrataClone Ultra PCR cloning system, which includes the Pfu Ultra II Fusion Hotstart DNA Polymerase.
StrataClone PCR Cloning Kit: High efficiency UA PCR cloning featuring DNA topoisomerase I and Cre recombinase technologies
StrataClone Blunt PCR Cloning Kit: High efficiency blunt-end PCR cloning, features DNA topoisomerase I and Cre recombinase technologies.
StrataClone Ultra Blunt PCR Cloning Kit: High efficiency blunt-end PCR cloning, features DNA topoisomerase I and Cre recombinase technologies, Kit includes PfuUltra II for high fidelity PCR amplification.
Caution: For research use only. Not for use in diagnostic procedures.