InterPlay Adenoviral TAP System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
240106 240101 240102
CA95040-754EA 182.37 CAD
CA95040-754 CA99900-120 CA99900-228 CA99900-962 CA99900-964 CA99900-966
InterPlay Adenoviral TAP System, Agilent Technologies
Vectors, Plasmids and Libraries
The InterPlay Mammalian TAP system allows you to recover interacting proteins from mammalian cells.
  • Protease digestion unnecessary to recover intact interacting protein partners since SBP and CBP elute easily from their affinity resins
  • Exceptionally pure protein for downstream analysis
This method is based on expression of a protein of interest fused to two affinity tags: a streptavidin binding peptide (SBP) and a calmodulin binding peptide (CBP). Tandem affinity purification yields your tagged protein and interacting proteins using gentle washing and small molecule elution conditions.
Caution: For research use only. Not for use in diagnostic procedures.