Glogos II Autorad Markers, Agilent Technologies
Supplier: Agilent
420202 420201 420202 420203 420201 420203
CA95040-686EA 1017.64 CAD
95040-688 ST420201 CA95040-686 CA95040-688 CAST420201 95040-686
Glogos II Autorad Markers, Agilent Technologies
Protein Linking and Labeling Systems
Luminescent peel-off stickers are used for accurately marking and aligning autoradiographs.
  • Adaptable for long or short exposures
Easily affixed to plastic wrap or paper, these convenient markers emit low-level luminescence rather than radioactivity, making the Glogos II markers a safe, non-hazardous alternative to radioactive ink.