Brilliant II SYBR® Master Mix, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Brilliant II SYBR® QPCR and QRT-PCR reagents were developed for improved sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
- Earlier Ct detection
- Improved data reproducibility
- Greater flexibility for use with numerous different templates and targets
- More accurate quantification results
- Developed on the Agilent Mx QPCR systems
When compared to other commercially available QPCR reagents, we observe earlier threshold cycle (Ct) detection at high and low template concentrations as well as improved reaction efficiency and replicate reproducibility across the entire range of template concentrations.
Caution: For research use only. Not for use in diagnostic procedures.