Ammonium hydrogen carbonate ≥99% eluent additive, for LC-MS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Ammonium bicarbonate

40867-50G
BJ40867-50GEA 124.22 CAD
BJ40867-50G
Ammonium hydrogen carbonate ≥99% eluent additive, for LC-MS, Fluka™
Ammonium hydrogen carbonate

Used as a eluent additive for LC-MS.

Formula: NH₄HCO₃
MW: 79.06 g/mol
Melting Pt: 60 °C
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012138
CAS Number: 1066-33-7
EINECS: 213-911-5
Merck Index: 13,00497

Order Now

Specification Test Results

Assay 99.5 - 101.0 %
pH (1 M, 20°C) 7.0 - 8.5
Solution 1 M in H₂O complying
Aluminium (Al) Max. 5 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 50 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 500 ppm
Manganese (Mn) Max. 5 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Suitability for the LC-MS complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR