Sodium nitrite 0.1 M NaNO2 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Diazotizing salts
35273-1L
BJ35273-1LEA
111.42
CAD
BJ35273-1L
Sodium nitrite 0.1 M NaNO2 (0.1N) volumetric solution, Fluka™
Sodium nitrite
Formula:
NaNO₂ MW: 69 g/mol Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011118 CAS Number: 7632-00-0 |
Specification Test Results
Factor | 0.998 - 1.002 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...