Tryptic Soy Broth (TSB), Teknova
Supplier: Teknova
T0420 T0430 T0430 T0425 T0420 T0425
CA101320-640EA 9474.15 CAD
101320-640 CA101320-640 101320-638 101320-636 CA101320-638 CA101320-636
Tryptic Soy Broth (TSB), Teknova
Media for Microbiology Dehydrated Media for Microbiology
TSA is a general purpose media produced via enzymatic digestion of soybean meal and casein. TSA is frequently the base media of other agar types; for example, blood agar plates are made by enriching TSA plates with blood.
TSA plates support growth of many semi-fastidious bacteria, including some species of Brucella, Corynebacterium, Listeria, Neisseria, and Vibrio.
1.5% Tryptone, 0.5% Soytone, 0.5% Sodium Chloride, 1.5% Agar.