2-Hexyl-1-decanol ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Guerbet hexadecanol, Hexyldecanol, 2-Hexyldecanol

H1461-25ML H1461-500ML H1461-100ML
TCH1461-25MLEA 69.99 CAD
TCH1461-25ML TCH1461-500ML TCH1461-100ML
2-Hexyl-1-decanol ≥97.0% (by GC)
2-Hexyl-1-decanol
Formula: C₁₆H₃₄O
Boiling Pt: 107 °C (0.1 mmHg) (lit.)
Melting Pt: –15 °C (lit.)
Flash Pt: 159 °C (lit.)
MDL Number: MFCD00060903
CAS Number: 2425-77-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12dd165bf36e55424aac2b243e35aa87e1aec68b&vid=12dd165bf36e55424aac2b243e35aa87e1aec68b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) Min. 97.0%
Specific gravity (20/20) 0.8370 - 0.8410
Refractive index n20/D 1.4470 - 1.4510


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR