KIMAX® Funnels, Long Stem, Thistle Top, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
46188-65300 46188-65400
16143-176EA 73.02 CAD
16143-176 16143-177
KIMAX® Funnels, Long Stem, Thistle Top, Kimble Chase
Funnels Filtration Funnels Filter Funnels
Funnels are manufactured from low extractable borosilicate glass.
  • Funnels are cylindrical in shape
  • 12" long stem, thistle top funnels
Certifications: Designed from USP Type I and ASTM E438, Type I, Class A requirements.