Nonsterile New Sponge Dressings, DUKAL™ Corporation
Supplier: Dukal
6112 6113 6114
95041-754CS 380.07 CAD
95041-754 95041-758 95041-762 CA95041-754 CA95041-758 CA95041-762
Nonsterile New Sponge Dressings, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Nonsterile new sponges are a rayon/poly blend construction that are virtually non-linting.
  • Rayon/poly blend construction
  • The effective combination of weight and open aperture allows each sponge to wick quicker and offer superior absorbency
  • Virtually non-linting
  • Not made with natural rubber latex
Order Now
SPECIFICATIONS
Environmentally Preferable