DuPont™ Tyvek® IsoClean® Boot Covers with PVC Sole, Elastic Ankles
Supplier: DuPont
IC444SWHLG02000B IC444SWHXL02000B
CA13452-164CS 3050.53 CAD
CA13452-164 CA13452-166
DuPont™ Tyvek® IsoClean® Boot Covers with PVC Sole, Elastic Ankles
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • PVC soles provide skid-resistance and durability
  • Elastic at ankles allows for a secure fit
  • Elastic opening
  • Elastic at ankles
  • Coverage up to calf
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
Packaging: Bulk packagedMore Product Information