Sulfur Dioxide Apparatus, General Glass Blowing
Supplier: General Glass Blowing
GW600 GW610 GW620 GW630 GW640 GW650 GW660 GW670
89107-100EA 2202.14 CAD
89107-100 89107-102 89107-104 89107-106 89107-108 89107-110 89107-112 89107-114
Sulfur Dioxide Apparatus, General Glass Blowing
Distillation Components Distillation Sets
The apparatus incorporates aeration-oxidation (Vacuum aspiration) method, in which a stream of air is passed through an acidified wine sample carrying released SO₂ through a condenser into a hydrogen peroxide trapping solution containing acid/base indicator. Precision of the analysis is dependent largely upon thorough consistent aeration and subsequent efficient trapping in the H₂O₂ solution.
The improved apparatus replaces the inefficient and imprecise Pasteur pipet sparging and impinger tubes with precise glass frits to assure thorough sparging the binding of SO. In addition, the use of glass frits results in an apparatus much less sensitive to small changes in the aspiration rate.