, anhydrous ≥98.0%, EMPROVE® ESSENTIAL E340, BP, Ph. Eur.

Supplier: EMD MILLIPORE – PCS CA EXCEP

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

1.05101.9029 1.05101.9525
CA1.05101.9029EA 2829.26 CAD
CA1.05101.9029 CA1.05101.5000 CA1.05101.9525
, anhydrous ≥98.0%, EMPROVE® ESSENTIAL E340, BP, Ph. Eur.
di-Potassium hydrogen phosphate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Corresponds to Ph. Eur., BP, E 340


Conforms to the purity criteria on food additives according to the current European Commission Regulation.

Formula: K₂HPO₄
Melting Pt: >450 °C Dec.
Density: 2.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (alkalimetric, calculated on dried substance) 98.0 - 101.0 %
Assay (out of dried substance) ≥ 98.0 %
Identity passes test
Appearance of solution passes test
In water insoluble matter ≤ 0.2 %
pH-value (1 %, water) 8.7 - 9.4
Chloride (Cl) ≤ 0.02 %
Fluoride (F) ≤ 0.0010 %
Sulfate (SO₄) ≤ 0.1 %
Heavy metals (as Pb) ≤ 0.0010 %
Al (Aluminium) ≤ 0.0005 %
As (Arsenic) ≤ 0.0001 %
Cd (Cadmium) ≤ 0.0001 %
Fe (Iron) ≤ 0.0010 %
Hg (Mercury) ≤ 0.0001 %
Na (Sodium) ≤ 1.5 %
Pb (Lead) ≤ 0.0001 %
Potassium dihydrogen phosphate ≤ 2.5 %
Residual solvents (ICH Q3C) excluded by manufacturing process
Reducing substances passes test
Loss on drying (130 °C) ≤ 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR