Digi-Sense® Advanced Pre-Calibrated Thermocouple Thermometers, Cole-Parmer
Supplier: Antyila Scientific
20250-91 20250-92
76183-384EA 356.51 CAD
76183-384 76183-386
Digi-Sense® Advanced Pre-Calibrated Thermocouple Thermometers, Cole-Parmer
Thermometers Digital Thermometers
Perfect for the lab to the production floor.
  • Reliable measurements guaranteed
  • NIST-traceable certificate included
  • Heavy-duty construction to protect against knocks and jarring
  • Super-large, dual, backlit LCD for easy data viewing
  • Accepts a wide variety of probes
  • Min., Max., and Avg. readings for quick object analysis
  • Electronic Offset function allows compensation of probe errors
  • Data logging model offers 16,000 data point memory and alarm
The Digi-Sense Professional Thermocouple Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours.
Digi-Sense instrument are fully tested and calibrated to NIST-traceable standards, saving you extra costs and waiting time to calibrate separately.
Certifications: NIST-traceable calibration certificate with data, performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005.
Delivery information: Includes protective rubber armor boot with built-in stand, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. Data logging model 20250-92 also includes a USB cable and software.