Digi-Sense® Pre-calibrated CFM/CMM Hot-Wire Thermo Anemometer, Cole-Parmer
Supplier: Cole Parmer
Digi-Sense®
2025016
76183-370EA 660.27 CAD
76183-370
Digi-Sense® Pre-calibrated CFM/CMM Hot-Wire Thermo Anemometer, Cole-Parmer
Anemometers
High precision at very low air velocities and flow rates.
  • Reliable measurements guaranteed
  • Pre-calibrated to NIST-traceable standards
  • Measure air velocities up to 4925 ft./min. (25 m/s) to cover a broad range of applications
  • Measure airflow up to 99,900 CFM (99,900 CMM)
  • Large, dual-function LCD with backlight capability for easy reading
  • Auto power-off extends battery life
The Digi-Sense hot-wire thermoanemometer is designed for precision air velocity and airflow measurements in seven user-selectable units (ft/min, m/s, mph, km/h, knots, CFM, or CMM). The instrument will help you meet cooling and environmental requirements for safety and comfort, as well as assure proper ventilation and filtration in cleanrooms and fume hoods.
The large dual display allows you to simultaneously read air velocity or airflow and air temperature and shows real time, maximum, minimum, and average values for the parameters under investigation. Slim telescopic probe is designed to reach into small openings.
The handheld hot-wire anemometer permits single point measurements or average measurements taken over a given time period. Comes in a foam-cut hard carrying case to protect the instrument between uses. Includes software and USB cable to download measurements into a PC for analysis and storage. Low-battery indicator prompts battery replacement, preventing power loss.
Certifications: NIST-traceable calibration certificate with data, performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005.
Delivery information: Includes telescoping hot-wire probe sensor with cable, USB cable, software CD, hard carrying case, AC/DC power adapter, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal.