Basic Blue 24 (New Methylene Blue) ≥90% (dye content) C.I. 52030
Supplier: CHEM-IMPEX INTERNATIONAL, INC
Synonyms:
New Methylene Blue N, zinc chloride double salt, New Methylene Blue N
22927-25G
22927-5G
CH22927-25GEA
308.1
CAD
CH22927-25G
CH22927-5G
Basic Blue 24 (New Methylene Blue) ≥90% (dye content) C.I. 52030
Basic Blue 24 (New Methylene Blue)
Formula:
C₁₈H₂₂ClN₃S·0.5 ZnCl₂ MW: 416.05 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00078135 CAS Number: 6586-05-6 |
Specification Test Results
Purity (Dye Content) | ≥90% |
Loss on Drying | ≤5% |
Spectro Photometric Assay | 5 mg/L in Water |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...