Industrial Gloves, MaxiFlex® Ultimate™ 34-874, Protective Industrial Products
Supplier: PIP
34-874/L 34-874/XL 34-874/M 34-874/S
20002-352CS 3230.61 CAD
20002-352 20002-354 20002-356 20002-358
Industrial Gloves, MaxiFlex® Ultimate™ 34-874, Protective Industrial Products
Gloves
The MaxiFlex® Ultimate™ has become the benchmark for precision handling in dry environments.
  • Soft liner made from Nylon and Spandex
  • Micro-foam NBR (Nitrile Butadiene Rubber) palm coating for controlled grip
  • Silicone free
  • FDA compliant, Food contact approved (EU)
  • Touchscreen compatible
The patented micro foam nitrile coating offers 360° breathability, reduces hand fatigue and increases comfort. Gloves are washable.
MaxiFlex® has outstanding durability withstanding 18.000 abrasive cycles per millimetre thanks to the use of the DURAtech® coatingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Ambidextrous/Hand-specific Hand-specific
Disposable / reusable Reusable
Liner material Nylon/Spandex
Finish Palm coating
Cuff style Knit wrist
FDA regulated +
Food contact approved +