WFI Sampler, Avantor Fluid Handling
Supplier: AVANTOR FLUID HANDLING, LLC
TS4-1200-AT-018 TS4-1200-AT-036 TS4-1600-AT-018 TS4-1600-AT-036
76175-268CS 4494.1 CAD
76175-268 76175-270 76175-272 76175-274
WFI Sampler, Avantor Fluid Handling
Samplers Fluid Samplers
WFI Samplers are an easy and convenient solution for obtaining perfect fluid samples from a WFI water drop.
  • Class VI Compliant Materials: made from AdvantaFlex tubing
  • Polypropylene fitting is inert and strong
  • Silicone tubing and gasket: unlike TPE materials, silicone will not contribute to TOC counts
  • Gamma-irradiation (SAL of 10–6) eliminates additional bio-burden concerns
  • Certified free from any animal derived components or processes
Each WFI Sampler is double-polypouched for cleanroom use, and irradiated. The fittings utilize the SterilEnz-II/ATMore Product Information