Digi-Sense® 400 Series Thermistor Probes with Handles, Cole-Parmer
Supplier: Antyila Scientific
93823-05 93824-00 93824-30
76183-380EA 124.92 CAD
76183-380 76183-390 76183-392
Digi-Sense® 400 Series Thermistor Probes with Handles, Cole-Parmer
Temperature Probes
Thermistor Probes offer excellent accuracy over the biological temperature range.
  • Probes are electronically isolated and with a 1/4" mono plug (unless otherwise specified)
  • Probes are designed for multiple applications
  • Time constant: 1 to 4 seconds or greater
Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five.
Note: The temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104 °C); PVC handles are rated to 200 °F (93 °C); Stainless Steel handles are rated at 450 °F (232 °C); Mini Connectors are rated to 392 °F (200 °C).