Digi-Sense® Thermocouple General-Purpose Surface Probes, Cole-Parmer
Supplier: Antyila Scientific
08516-60 08517-60 08500-60
76183-442EA 204.72 CAD
76183-442 76183-450 76183-466
Digi-Sense® Thermocouple General-Purpose Surface Probes, Cole-Parmer
Temperature Probes
Designed to measure the temperature at the surface without penetrating the material.
  • These probes are designed to measure surface temperature without penetrating the surface
  • Ideal for monitoring flat surfaces such as hot plates, furnaces, or molds
  • Response Time: 1.25 seconds or greater
  • Each probe comes with color-coded ANSI coiled cable: Type J-black, Type K-yellow, Type T-blue, and Type E-purple
  • Custom designs or changes to this product are available upon request
Temperature accuracy for the Type–T probe is ±3.6 °F (±2.0 °C), or ±0.4% of reading above 32 °F (0 °C); ±0.8% of reading below 32 °F (0 °C). Temperature accuracy for the Type–K probe is ±3.0 °F (1.1 °C) or ±0.4% reading above 212 °F(100 °C); ±3.6 °F (2.2 °C) or 2% of reading between –58 to 211 °F (–50 to 99 °C); ±7.2 °F (4 °C) or ±4% of reading between –60 to –238 (–51 to –150 °C), whichever is greater. Temperature accuracy for the Type–J probe is ±5.9 °F (±3.3 °C), or ±0.4% of reading above 32 °F (0 °C); ±4.0% of reading below 32 °F (0 °C).