Straight Sided Stoppers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
73811T-13 73811T-21 73811-21
66004-000CS 244.11 CAD
66004-000 66004-002 66004-004 66004-006
Straight Sided Stoppers, DWK Life Sciences
Stoppers Rubber Stoppers
These high quality gray chlorobutyl stoppers are used for research and pharmaceutical packaging applications.
  • Formulation contains no plasticizers, 2-mercapto-benzothaizole, nitrosamine precursors or natural rubber latex
  • Formulation is applicable to aqueous solutions with a pH range of 2 to 10
  • Durometer 50
  • Universal gray chlorobutyl formulation passes Japanese, European and United States pharmacopeia testing for globally marketed pharmaceutical products
*Is rinsed with a silicon oil to provide increased lubricity.
Order Now
SPECIFICATIONS
Environmentally Preferable