Digi-Sense® Thermocouple General-Purpose Penetration Probes, Cole-Parmer
Supplier: Antyila Scientific
08517-65 08516-65 08500-65
76183-354EA 167.93 CAD
76183-354 76183-444 76183-588
Digi-Sense® Thermocouple General-Purpose Penetration Probes, Cole-Parmer
Temperature Probes
General-purpose penetration probes are designed for soft and semi-soft materials.
  • Response time: 2 seconds or greater
  • ABS handles offer a lightweight design for ease of use, along with durability
  • Probe handles feature a strain relief spring to protect the cable connection against damage due to repeated flexing and tugging
  • Each probe comes with color-coded ANSI mini connectors: Type J-black, Type K-yellow, and Type T-blue
  • Custom designs or changes to this product are available upon request
Temperature accuracy for the Type–T probe is ±3.6 °F (±2.0 °C), or ±0.4% of reading above 32 °F (0 °C); ±0.8% of reading below 32 °F (0 °C). Temperature accuracy for the Type–K probe is ±3.0 °F (1.1 °C) or ±0.4% reading above 212 °F(100 °C); ±3.6 °F (2.2 °C) or 2% of reading between –58 to 211 °F (–50 to 99 °C); ±7.2 °F (4 °C) or ±4% of reading between –60 to –238 (–51 to –150 °C), whichever is greater. Temperature accuracy for the Type–J probe is ±5.9 °F (±3.3 °C), or ±0.4% of reading above 32 °F (0 °C); ±4.0% of reading below 32 °F (0 °C).