Digi-Sense® Thermocouple General-Purpose Probes with Mini connector, Cole-Parmer
Supplier: Antyila Scientific
08516-55 08517-55 08500-55
76183-440EA 148.14 CAD
76183-440 76183-448 76183-464
Digi-Sense® Thermocouple General-Purpose Probes with Mini connector, Cole-Parmer
Temperature Probes
General-purpose probes are designed for use with liquids, gases, and semisolids.
  • Response time: 1.7 seconds or greater
  • ABS handles offer a lightweight design for ease of use, along with durability
  • Probe handles feature a strain relief spring to protect the cable connection against damage due to repeated flexing and tugging
  • Each probe comes with color-coded ANSI mini connectors: Type J-black, Type K-yellow, Type T-blue, and Type E-purple
  • Custom designs or changes to this product are available upon request
Temperature accuracy for the Type–T probe is ±3.More Product Information