Digi-Sense® RTD Probe with Handle, Cole-Parmer
Supplier: Antyila Scientific
08117-70 08117-75 08117-85 08117-90
76183-456EA 385.67 CAD
76183-456 76183-458 76183-460 76183-462
Digi-Sense® RTD Probe with Handle, Cole-Parmer
Temperature Probes
Designed to provide the same accuracy and benefits of an RTD Probe in a compact and economical design.
  • Time constant: 1–3 seconds or greater
  • Maximum error for probe assembly is ±1.1°C + 0.12% of reading below 300 °F, ±0.35% of reading above 300 °F
  • Alpha coefficient is 0.003850 Ω/Ω/°C (DIN IEC 751)
  • Resistance at ice point (0°C) is 100 Ω
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).