DuPont™ Tyvek® IsoClean® Boot Covers
Supplier: DuPont
IC458BWHLG01000B IC458BWHXL01000B IC458BWHMD01000B IC458BWHLG0100CS IC458BWHMD0100CS IC458BWHXL0100CS IC458BWHLG0100CS IC458BWHSM0100CS
CA89125-640CS 2994.37 CAD
CA89125-640 CA89125-642 CA89127-338 CA89127-336 CA89127-340 CA89129-686 89127-336 10016-100
DuPont™ Tyvek® IsoClean® Boot Covers
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Elastic opening
  • Ankle ties allow for a secure fit
  • Coverage up to calf
Made using a flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged option packed in double transparent poly liners.