Trio.Bas™ Gas System with MINI Instrument Kit
Supplier: HARDY DIAGNOSTICS CA
BAS654K BAS655K
CA76196-438EA 39834.75 CAD
CA76196-438 CA76196-440
Trio.Bas™ Gas System with MINI Instrument Kit
Samplers Air Samplers
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
  • Aspirating head with easy manipulation bayonet closure
  • Manual or automatic operations
  • Ergonomic and balanced design to facilitate handling with ease
  • The air sampler is easily connected to the output of the gas system
  • Each component of the Trio.Gas™ adapter instrument is autoclavable
  • Calibrated regulator guarantees a 100 liters per minute flow rate
The Trio.Bas™ instruments utilize the impaction method of air sampling. The air, passing through an aspirating head, impacts onto an appropriate agar plate that collects bacteria or other organic substances possibly present in the sampled area. All air samplers are lightweight, with an antibacterial techno-polymer shockproof body. They have a long lasting battery and are IP65 protected from dust and water.
The Trio.Gas™ microbial compressed gas system is designed to test the presence of microorganisms in compressed gas samples supplied from tanks and pipes under pressure.
Certifications: According to ISO14698, USP 797 (Impaction is the preferred method of active air sampling. Use of settling plates for qualitative air sampling cannot be relied upon and shall not be used solely to determine the quality of air in the controlled environment), USP 116, EU GPL-GMP, ISO17025.
Accessories information: Additional accessory components, including data transfer options, are available separately.
Ordering information: If automatically timed samples and electronic data records are required, the MINI or MONO instrument used concurrently with the Gas system is necessary.
If manually timed samples and hand recorded sampling data is acceptable, the MINI or MONO instrument is not necessary. Purchase the Gas system 76196-434 or 76196-436, which includes gas chambers. Choose kit based on plate size preference.
Delivery information: Trio.Bas™ Gas System with MINI Instrument Kit Contains:
Trio.Gas™ System, high pressure gas sampler attachment, includes stainless steel electro valve and fixing system for air sampler, and gas connection
Trio.Bas™ MINI: Includes calibration certificate, 100 L/min. flow rate, Bluetooth data transfer capability (choose desired data download method, separate purchase required), Contact or Petri plate variations
Battery wall charger with cable, 100/240V battery
Stainless steel aspirating head and protection cover head, autoclavable
IQ, OQ, PQ, fillable documents