DuPont™ Tyvek® 400 Coveralls with Open Wrists and Ankles, Snap Front Closure, Comfort Fit Design
Supplier: DuPont
TY151SWH2X002500 TY151SWH3X002500 TY151SWH4X002500 TY151SWHLG002500 TY151SWHMD002500 TY151SWHXL002500
10016-702CS 427.31 CAD
10016-702 10016-704 10016-706 10016-708 10016-710 10016-712
DuPont™ Tyvek® 400 Coveralls with Open Wrists and Ankles, Snap Front Closure, Comfort Fit Design
Disposable Garments
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
  • Comfort fit design
  • Laydown collar
  • Snap front
  • Open wrists and ankles
  • Elastic waist
  • Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Comfort fit design is based on extensive wearer input, to provide our most comfortable garment design that enables a greater range of movement while stretching and bending, provides a more tailored fit, offers reinforcement in high stress areas for fewer blowouts, utilizes Snap front for easier donning and doffing and an elastic waist to better position the garment. Coveralls feature a laydown collar, snap front, and open wrists and ankles.
Certifications: Thickness meets ASTM D1777.