DuPont™ ProShield® 10 Coveralls with Standard Hood
Supplier: DuPont
ProShield®
PB127SBUSM002500 PB127SGYMD002500 PB127SWHSM002500 PB127SGY4X002500 PB127SBU4X002500 PB127SWH5X002500 PB127SWH2X002500 PB127SGYXL002500 PB127SWH3X002500 PB127SGY3X002500 PB127SGY6X002500 PB127SBULG002500 PB127SGY2X002500 PB127SBU5X002500 PB127SBUMD002500 PB127SWHLG002500 PB127SBU7X002500 PB127SWH6X002500 PB127SGY5X002500 PB127SWHMD002500 PB127SGY7X002500 PB127SWH7X002500 PB127SWH4X002500 PB127SBU3X002500 PB127SBU6X002500 PB127SGYLG002500 PB127SBU2X002500 PB127SBUXL002500 PB127SWHXL002500
10016-756CS 156.21 CAD
76187-492 76187-494 76187-496 76187-830 10016-756 10016-758 76187-826 10016-264 76187-828 10016-760 10016-266 10016-762 10016-268 10016-764 76187-490 10016-262 76187-486 76187-488 10016-766 10016-768 10016-748 10016-274 10016-770 10016-750 10016-772 10016-752 10016-754 10016-270 10016-272
DuPont™ ProShield® 10 Coveralls with Standard Hood
Disposable Garments
DuPont™ ProShield® 10, spun bond melt blown spun bond (SMS) garments, are made to the high-quality standards you have come to expect from DuPont.
  • Standard fit hood
  • Elastic wrists and ankles
  • Elastic waist
  • Serged seams
ProShield® 10 garments have been optimized for comfort, softness and breathability while enabling you to cut costs for disposable garments.
DuPont™ ProShield® 10 disposable garments are designed for workers in a range of industries for nonhazardous dry particle and light liquid splash applications. Garments are available in a variety of coverall styles in a choice of white, blue and gray. Applications include: general maintenance, janitorial/cleaning and other dirty work.
Attached hood features elastic around the face opening. Elastic openings and wrists and ankles allows for a tighter fit.
Certifications: Thickness meets ASTM D1777.