Prostaglandin F ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
516011-480 516011-96
75816-884EA 2497.27 CAD
75816-884 75816-886
Prostaglandin F ELISA Kit, Cayman Chemical Company
Assays ELISAs
For the quantification of PGF in tissue culture supernatants and other sample matrices.
  • Measure PGF in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure PGF levels down to 10 pg/mL
  • Incubation: 18 hours | Development: 90 – 120 minutes | Read: Colorimetric at 405 – 420 nm
PGF is one of the five primary PGs derived enzymatically directly from the endoperoxide PGH2. The majority of the functional roles ascribed to it relate to fertility, pregnancy, and parturition. Like all of the primary PGs, PGF has a very short half-life in the general circulation. The plasma concentration of PGF in humans is <10 pg/mL, and probably no more than 1 – 2 pg/mL. Cayman’s PGF ELISA Kit is a competitive assay that can be used for the quantification of PGF in tissue culture supernatants and other sample matrices.