Anti-CGB Mouse Monoclonal Antibody (CF647) [clone: HCGb/54 HCGb/459]

Supplier: Biotium

BNC471224-100 BNC471224-500
75951-824EA 325.12 CAD
75951-824 75951-826
Anti-CGB Mouse Monoclonal Antibody (CF647) [clone: HCGb/54 HCGb/459]
Antibodies
This MAb reacts with a protein of 22 kDa, identified as β sub-unit of HCG. It does not cross react with the α sub-unit. HCG is a glycoprotein, which is secreted in large quantities by normal trophoblasts. It is present only in trace amounts in non-pregnant urine and sera but rises sharply during pregnancy. HCG is composed of two non-identical, non-covalently linked polypeptide chains designated as the alpha and beta subunits. The beta subunit is identical to that of thyroid stimulating hormone (TSH), follicle stimulating hormone (FSH), and luteinizing hormone (LH). hCG MAb detects cells and tumors of trophoblastic origin such as choriocarcinoma. Large cell carcinoma and adenocarcinoma of the lung demonstrate anti-hCG positivity in 90% and 60% of cases respectively. 20% of lung squamous cell carcinomas are positive. hCG expression by non-trophoblastic tumors may indicate aggressive behavior.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: CGB
Clonality: Monoclonal
Clone: Clone HCGb/54 HCGb/459
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG's
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR