Anti-CD74 Mouse Monoclonal Antibody (CF568) [clone: LN-2 CLIP/813]

Supplier: Biotium

BNC681207-100 BNC681207-500
75907-438EA 325.12 CAD
75907-438 75907-440
Anti-CD74 Mouse Monoclonal Antibody (CF568) [clone: LN-2 CLIP/813]
Antibodies
This antibody recognizes a protein of ~35 kDa, identified as CD74. It is a type II transmembrane protein which binds to the peptide binding groove of newly synthesized MHC class II alpha/beta heterodimers and prevents their premature association with endogenous polypeptides. CD74 is expressed primarily by antigen presenting cells, such as B-lymphocytes (from before the pre-B cell stage to before the plasma cell stage), macrophages, and monocytes, and many epithelial cells. Anti-CD74 stains predominantly germinal center lymphocytes and B-cell lymphomas, but rarely T-cell lymphomas. Anti-CD74More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD74
Antigen name CD74
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone LN-2 CLIP/813
Reactivity Human
Host Mouse
Gene ID Unigene ID: 436568
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR