Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: KRT7/760 OV-TL12/30]

Supplier: Biotium

BNC401167-100 BNC401167-500
75920-048EA 325.12 CAD
75920-048 75920-050
Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: KRT7/760 OV-TL12/30]
Antibodies
This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: Clone KRT7/760 OV-TL12/30
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR