Anti-KRT76 Mouse Monoclonal Antibody (CF568) [clone: KRTH/1076]

Supplier: Biotium

BNC681076-100 BNC681076-500
75915-932EA 325.12 CAD
75915-932 75915-934
Anti-KRT76 Mouse Monoclonal Antibody (CF568) [clone: KRTH/1076]
Antibodies
This MAb recognizes basic (Type II or HMW) cytokeratins, which include 67 kDa (CK1); 64 kDa (CK3); 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8). Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50 , 48, 46, 45, and 40 kDa. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightnessMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol KRT76
Environmentally Preferable
Antigen name Cytokeratin HMW
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone KRTH/1076
Reactivity Human,Rat
Host Mouse
Gene ID Unigene ID: 654392
Isotype IgG1 kappa
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR