Anti-SOX10 Mouse Monoclonal Antibody (Biotin) [clone: SOX10/991 SOX10/1074]

Supplier: Biotium

BNCB1074-100 BNCB1074-500
75973-488EA 325.12 CAD
75973-488 75973-490
Anti-SOX10 Mouse Monoclonal Antibody (Biotin) [clone: SOX10/991 SOX10/1074]
Antibodies
This antibody recognizes a protein of ~55 kDa, identified as SOX10. This MAb is highly specific and does not cross-react with other members of the SOX-family. SOX genes comprise a family of genes that are related to the mammalian sex-determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. SOX-10 is a sensitive marker of melanoma, including conventional, spindled, and desmoplastic subtypes. It is expressed by metastatic melanomas and nodal capsular nevus in sentinel lymph nodes, but not by other lymph node components such as dendritic cells, which usually express S100 protein. Commonly used melanoma markers, such as anti-HMB-45 and anti-Melan-A, are poorly expressed in desmoplastic melanomas while SOX-10 is moderately to strongly expressed in desmoplastic melanomas. SOX-10 is considered as a very reliable marker for recognizing residual desmoplastic melanomas. In normal tissues, it is expressed in Schwann cells, melanocytes, and myoepithelial cells of salivary, bronchial and mammary glands. SOX-10 expression is also observed in mast cells.

Type: Primary
Antigen: SOX10
Clonality: Monoclonal
Clone: Clone SOX10/991 SOX10/1074
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR