Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF647) [clone: HLA-DRB/1067]

Supplier: Biotium

BNC471067-100 BNC471067-500
75951-436EA 325.12 CAD
75951-436 75951-438
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF647) [clone: HLA-DRB/1067]
Antibodies
This MAb reacts with the beta-chain of HLA-DRB1 antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. Its epitope is different from that of MAb L243. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36 kDa alpha (heavy) chain and a 28 kDa beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. It is an excellent histiocyticMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol HLA-DRB1
Antigen name HLA-DRB
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 647
Clone HLA-DRB/1067
Reactivity Monkey,Human
Host Mouse
Gene ID Unigene ID: 534322
Isotype IgG2b kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR