Anti-FSHB Mouse Monoclonal Antibody (CF640R) [clone: FSHb/1062]

Supplier: Biotium

BNC401062-100 BNC401062-500
75926-756EA 325.12 CAD
75926-756 75926-758
Anti-FSHB Mouse Monoclonal Antibody (CF640R) [clone: FSHb/1062]
Antibodies
This MAb reacts with a protein of 22 kDa, identified as beta sub-unit of FSH. It does not cross react with the alpha sub-unit. Follicle stimulating hormone (FSH) is a hormone synthesized and secreted by gonadotrophs in the anterior pituitary gland. In the ovary, FSH stimulates the growth of immature Graafian follicles to maturation. As the follicle grows, it releases inhibin, which deactivates the FSH production. In men, FSH enhances the production of androgen-binding protein by the Sertoli cells of the testis and is critical for spermatogenesis. FSH and LH act synergistically in reproduction. FSH is a useful marker in the classification of pituitary tumors and the study of pituitary disease.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: FSHB
Clonality: Monoclonal
Clone: FSHb/1062
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR