Anti-CHGA Mouse Monoclonal Antibody (CF640R) [clone: CGA414 CGA/777 CGA/798]

Supplier: Biotium

BNC401060-100 BNC401060-500
75908-792EA 325.12 CAD
75908-792 75908-794
Anti-CHGA Mouse Monoclonal Antibody (CF640R) [clone: CGA414 CGA/777 CGA/798]
Antibodies
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raiseMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CHGA
Antigen name Chromogranin A
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 640R
Clone CGA414 CGA/777 CGA/798
Reactivity Pig,Monkey,Human,Rat,Mouse
Host Mouse
Gene ID Unigene ID: 150793
Isotype IgGs
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/ml
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR