Anti-CD48 Mouse Monoclonal Antibody [clone: 5-4.8]

Supplier: Biotium

BNUB1057-100 BNUB1057-500
75913-928EA 380.65 CAD
75913-928 75913-930
Anti-CD48 Mouse Monoclonal Antibody [clone: 5-4.8]
Antibodies
This antibody recognizes human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of the markers for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: Clone 5-4.8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR