Anti-CD48 Mouse Monoclonal Antibody (CF568) [clone: 5-4.8]

Supplier: Biotium

BNC681057-100 BNC681057-500
75913-962EA 325.12 CAD
75913-962 75913-964
Anti-CD48 Mouse Monoclonal Antibody (CF568) [clone: 5-4.8]
Antibodies
This antibody recognizes human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of the markers for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: Clone 5-4.8
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR