Anti-CD48 Mouse Monoclonal Antibody (CF640R) [clone: 5-4.8]

Supplier: Biotium

BNC401057-100 BNC401057-500
75913-970EA 325.12 CAD
75913-970 75913-972
Anti-CD48 Mouse Monoclonal Antibody (CF640R) [clone: 5-4.8]
Antibodies
This antibody recognizes human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of the markers for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: Clone 5-4.8
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR