Anti-FCGR3A Mouse Monoclonal Antibody (CF647) [clone: HO-80]
Supplier: Biotium
BNC471056-100 BNC471056-500
75888-506EA 325.12 CAD
75888-506 75888-508
Anti-FCGR3A Mouse Monoclonal Antibody (CF647) [clone: HO-80]
Antibodies
This antibody recognizes CD16 (FcγRIII), the low-affinity receptor for IgG with an apparent molecular weight of 50-80 kDa. Two similar genes represent CD16, CD16A (FcγRIIIA), which exists as a hetero-oligomeric polypeptide-anchored form in macrophages and NK cells and CD16B (FcγRIIIB), which exist as a monomeric GPI-anchored form in neutrophils. Furthermore, there are two known polymorphisms of CD16B, NA-1 and NA-2. Individuals homozygous for NA-2 show a lower phagocytic capacity compared with NA-1. CD16 binds IgG in the form of immune complexes and shows preferential binding of IgG1 and IgG3More Product Information