Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF405S) [clone: 246-B8.E7]

Supplier: Biotium

BNC041054-100 BNC041054-500
75960-856EA 325.12 CAD
75960-856 75960-858
Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF405S) [clone: 246-B8.E7]
Antibodies
This antibody recognizes a monomorphic determinant of human major histocompatibility (MHC) class I antigens (HLA-A, B and C). Human MHC class I antigens are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm)More Product Information
Order Now

SPECIFICATIONS

Antigen symbol HLA-A; HLA-B; HLA-C
Environmentally Preferable
Antigen name MHC I
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone 246-B8.E7
Reactivity Non-human primates,Human,Mouse
Host Mouse
Gene ID Unigene ID: 181244; 654404; 77961
Isotype IgG2a kappa
ImmunoFluorescence Yes
Concentration 0.1 mg/ml
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR