Anti-CD3E Mouse Monoclonal Antibody (Biotin) [clone: B-B12]

Supplier: Biotium

BNCB1052-100 BNCB1052-500
75895-354EA 325.12 CAD
75895-354 75895-356
Anti-CD3E Mouse Monoclonal Antibody (Biotin) [clone: B-B12]
Antibodies
This antibody recognizes five invariable CD3 chains (designated as and ) with molecular weight ranging from 16-28 kDa. CD3 is expressed, typically at high levels, on peripheral T cells and majority of T cell neoplasms. Thymocytes express CD3 at different level on the cell surface in the course of differentiation and, in cortical thymus, CD3 is predominantly Intracytoplasmic. The CD3 complex is closely associated at the lymphocyte cell surface with T cell antigen receptor (TCR) and is involved in transducing antigen-recognition signals into cytoplasm of T cells and in regulating the cell surface expression of the TCR complex.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: B-B12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR