Anti-B2M Mouse Monoclonal Antibody (CF568) [clone: 246-E9.E7]

Supplier: Biotium

BNC681039-100 BNC681039-500
75882-002EA 325.12 CAD
75882-002 75883-912
Anti-B2M Mouse Monoclonal Antibody (CF568) [clone: 246-E9.E7]
Antibodies
This antibody recognizes a protein of 12 kDa, identified asβ2-microglobulin.β2-microglobulin non-covalently associates with the 44 kDa chain to form the HLA Class I antigen complex. Human β2-microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human β2-microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of βMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol B2M
Environmentally Preferable
Antigen name Beta-2-Microglobulin
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone 246-E9.E7
Reactivity Human
Host Mouse
Gene ID Unigene ID: 534255
Isotype IgG2a kappa
ImmunoFluorescence Yes
Concentration 0.1 mg/ml
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR